Assay Details
Target Gene Details
Entrez Gene ID: | 8720 |
Gene Name: | membrane bound transcription factor peptidase, site 1 |
Gene Aliases: |
PCSK8, S1P, SKI-1 |
Location: |
Chr.16:84053761-84116943 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBTPS1 | NM_003791.3 | 16 | 2675 | NP_003782.1 |
XM_017023816.1 | 15 | 2424 | XP_016879305.1 | |
XM_017023817.1 | 17 | 2678 | XP_016879306.1 | |
AK091212.1 | 5 | 1385 | ||
AK291773.1 | 16 | 2558 | ||
AK299067.1 | 15 | 1900 | ||
BC114555.1 | 16 | 2561 | AAI14556.1 | |
BC114961.1 | 16 | 2561 | AAI14962.1 | |
BM048055.1 | ||||
D42053.1 | 16 | 2638 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639410 | Chr.16:84055831 - 84075312 on Build GRCh38 | Loss |
![]() |
esv3892924 | Chr.16:84014717 - 84091665 on Build GRCh38 | Gain |
![]() ![]() |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3892927 | Chr.16:84058861 - 84081674 on Build GRCh38 | Gain |
![]() |
nsv1909 | Chr.16:84031468 - 84076703 on Build GRCh38 | Deletion |
![]() ![]() |
dgv546e214 | Chr.16:84011032 - 84095132 on Build GRCh38 | Gain |
![]() ![]() |
nsv457595 | Chr.16:84013110 - 84091665 on Build GRCh38 | Gain |
![]() ![]() |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)