Assay Details
Target Gene Details
Entrez Gene ID: | 9625 |
Gene Name: | apoptosis associated tyrosine kinase |
Gene Aliases: |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
Location: |
Chr.17:81117295-81166072 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK | NM_001080395.2 | NP_001073864.2 | ||
XM_006722193.2 | XP_006722256.1 | |||
XM_006722194.2 | XP_006722257.1 | |||
AK131529.1 | BAD18667.1 |
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | MRPL12 MIR3186 LOC100129503 ARL16 TMEM105 MIR657 BAHCC1 CEP131 MIR4740 MIR3065 TEPSIN FSCN2 AATK LOC107985022 LOC100130370 ACTG1 LOC107985021 LOC101928855 CHMP6 SLC38A10 CCDC137 LOC400627 FAAP100 AATK-AS1 C17orf89 OXLD1 LOC105371921 HGS MIR1250 TSPAN10 BAIAP2 SLC25A10 NPLOC4 LOC105371925 MIR338 MIR6786 RPTOR BAIAP2-AS1 LINC00482 PDE6G |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | AATK AATK-AS1 CEP131 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MRPL12 UTS2R RAB40B ARL16 TMEM105 ASPSCR1 BAHCC1 SLC16A3 TEX19 FSCN2 RFNG LOC101929552 CCDC57 LOC105371944 FASN NOTUM SLC38A10 FN3K LOC101929511 AATK-AS1 MIR6787 HGS MAFG-AS1 ANAPC11 MIR4525 P4HB DCXR ALYREF OGFOD3 SLC25A10 NPLOC4 LOC105371925 RAC3 DUS1L SECTM1 LINC00482 MIR3186 PPP1R27 WDR45B FOXK2 ARHGDIA CEP131 PYCR1 MAFG MIR4740 GPS1 TEPSIN NARF FN3KRP HEXDC SIRT7 AATK LOC105376790 NPB TBCD LOC100130370 LRRC45 ACTG1 CCDC137 CD7 FAAP100 SNORD134 B3GNTL1 CSNK1D LOC105376791 C17orf89 MCRIP1 OXLD1 ZNF750 TSPAN10 MYADML2 C17orf62 STRA13 GCGR MIR6786 PCYT2 PDE6G |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | AATK AATK-AS1 LOC105371925 CEP131 C17orf89 TEPSIN SLC38A10 |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | MIR1250 AATK BAIAP2 AATK-AS1 MIR657 MIR338 CEP131 MIR3065 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | AATK-AS1 TMEM105 MIR657 CEP131 C17orf89 MIR3065 TEPSIN MIR1250 AATK BAIAP2 LOC105371925 MIR338 SLC38A10 BAIAP2-AS1 LINC00482 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR1250 AATK BAIAP2 AATK-AS1 MIR657 MIR338 CEP131 MIR3065 BAIAP2-AS1 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | FAAP100 MIR3186 AATK-AS1 TMEM105 BAHCC1 CEP131 C17orf89 MIR4740 TEPSIN FSCN2 AATK LOC100130370 ACTG1 NPLOC4 LOC105371925 SLC38A10 LINC00482 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | MIR1250 AATK BAIAP2 AATK-AS1 MIR657 MIR338 MIR3065 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |