Assay Details
Target Gene Details
Entrez Gene ID: | 669 |
Gene Name: | bisphosphoglycerate mutase |
Gene Aliases: |
DPGM |
Location: |
Chr.7:134646779-134679816 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BPGM | NM_001293085.1 | 3 | 381 | NP_001280014.1 |
NM_001724.4 | 2 | 256 | NP_001715.1 | |
NM_199186.2 | 3 | 577 | NP_954655.1 | |
XM_011516527.1 | 2 | 437 | XP_011514829.1 | |
AB209044.1 | 1 | 108 | BAD92281.1 | |
AK097778.1 | 2 | 200 | ||
AK315439.1 | 2 | 224 | ||
BC017050.1 | 2 | 227 | AAH17050.1 | |
BM090687.1 | 3 | 491 | ||
BU664387.1 | 3 | 510 | ||
BX473275.1 | 2 | 240 | ||
CB994648.1 | 3 | 533 | ||
CF242942.1 | 3 | 537 | ||
DA701935.1 | 3 | 349 | ||
DB047822.1 | 2 | 253 | ||
DB049554.1 | ||||
X04327.1 | 2 | 146 | CAA27858.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |