Assay Details
Target Gene Details
Entrez Gene ID: | 10495 |
Gene Name: | ecto-NOX disulfide-thiol exchanger 2 |
Gene Aliases: |
APK1, COVA1, tNOX |
Location: |
Chr.X:130622330-130903317 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENOX2 | NM_001281736.1 | 14 | 2096 | NP_001268665.1 |
NM_006375.3 | 15 | 2240 | NP_006366.2 | |
NM_182314.2 | 16 | 2329 | NP_872114.1 | |
XM_005262354.3 | 15 | 2185 | XP_005262411.1 | |
XM_011531244.2 | 16 | 2331 | XP_011529546.1 | |
XM_011531245.2 | 16 | 2325 | XP_011529547.1 | |
XM_011531247.2 | 17 | 2457 | XP_011529549.1 | |
XM_011531249.2 | 14 | 1885 | XP_011529551.1 | |
XM_011531251.2 | 14 | 1990 | XP_011529553.1 | |
XM_017029226.1 | 18 | 2548 | XP_016884715.1 | |
XM_017029227.1 | 16 | 2282 | XP_016884716.1 | |
XM_017029228.1 | 15 | 2031 | XP_016884717.1 | |
XM_017029229.1 | 15 | 2234 | XP_016884718.1 | |
AF207881.2 | 13 | 1850 | AAF20934.2 | |
AK000353.1 | 14 | 2013 | ||
AK289812.1 | 16 | 2221 | ||
AK289837.1 | 15 | 2132 | ||
BC019254.1 | 10 | 1365 | AAH19254.2 | |
BC139781.1 | 13 | 1913 | ||
BC140874.1 | 14 | 2010 | ||
S72904.1 | 8 | 1006 | AAB30428.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |