Assay Details
Target Gene Details
Entrez Gene ID: | 56916 |
Gene Name: | SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1 |
Gene Aliases: |
ADERM, ETL1, HEL1 |
Location: |
Chr.4:94207608-94291292 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMARCAD1 | NM_001128429.2 | 2 | 293 | NP_001121901.1 |
NM_001128430.1 | 2 | 195 | NP_001121902.1 | |
NM_020159.4 | 2 | 293 | NP_064544.2 | |
NR_045644.1 | 2 | 226 | ||
XM_017008463.1 | 2 | 117 | XP_016863952.1 | |
XM_017008464.1 | 2 | 117 | XP_016863953.1 | |
XM_017008465.1 | 2 | 127 | XP_016863954.1 | |
AB032948.2 | 1 | 81 | ||
AK027490.1 | 2 | 191 | BAB55150.1 | |
AL704040.1 | 2 | 294 | ||
AY008271.1 | 2 | 96 | AAG16639.1 | |
BC017953.1 | 2 | 178 | ||
BC045534.1 | 2 | 248 | AAH45534.1 | |
DA798320.1 | 2 | 195 | ||
DB207879.1 | 2 | 273 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |