Assay Details
Target Gene Details
Entrez Gene ID: | 6787 |
Gene Name: | NIMA related kinase 4 |
Gene Aliases: |
NRK2, STK2, pp12301 |
Location: |
Chr.3:52707936-52770949 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEK4 | NM_001193533.1 | 2 | 434 | NP_001180462.1 |
NM_003157.4 | 3 | 701 | NP_003148.2 | |
XM_006713310.2 | 3 | 698 | XP_006713373.1 | |
XM_011534039.2 | 3 | 698 | XP_011532341.1 | |
XM_011534040.2 | 3 | 698 | XP_011532342.1 | |
XM_017007085.1 | 3 | 698 | XP_016862574.1 | |
XM_017007086.1 | 3 | 698 | XP_016862575.1 | |
AK294165.1 | 2 | 341 | ||
AK312420.1 | 3 | 689 | ||
BC015515.1 | 3 | 665 | ||
BC063044.1 | 3 | 691 | AAH63044.1 | |
EF560744.1 | 3 | 697 | ||
KJ592714.1 | 3 | 498 | ||
L20321.1 | 3 | 676 | AAA36658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508219 | Chr.3:52707177 - 52769883 on Build GRCh38 | Deletion |
![]() ![]() |
esv2762328 | Chr.3:52740868 - 52968134 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834698 | Chr.3:52715967 - 52894203 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)