Assay Details
Target Gene Details
Entrez Gene ID: | 3299 |
Gene Name: | heat shock transcription factor 4 |
Gene Aliases: |
CTM, CTRCT5 |
Location: |
Chr.16:67163385-67169945 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSF4 | NM_001040667.2 | NP_001035757.1 | ||
NM_001538.3 | NP_001529.2 | |||
AB029348.1 | BAA84582.1 | |||
AK225152.1 | ||||
AK293631.1 | ||||
AK296210.1 | ||||
AK308638.1 | 3 | 1313 | ||
BM805441.1 | ||||
D87673.1 | BAA13433.1 | |||
DA372008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | EXOC3L1 TMEM208 NUTF2 LOC105369155 NOL3 FHOD1 ENKD1 GFOD2 TSNAXIP1 EDC4 ELMO3 ATP6V0D1 KIAA0895L CARMIL2 FAM65A PARD6A LRRC29 LOC107984813 ACD MIR328 THAP11 SLC12A4 NRN1L FBXL8 LRRC36 AGRP LCAT LOC100131303 C16orf86 HSD11B2 DDX28 TRADD ZDHHC1 RANBP10 LOC100505942 DUS2 DPEP2 CENPT CTCF PLEKHG4 DPEP3 PSKH1 HSF4 TPPP3 E2F4 SLC9A5 KCTD19 PSMB10 CTRL |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | KIAA0895L C16orf70 EXOC3L1 TMEM208 TRADD HSF4 E2F4 LOC105369155 SLC9A5 NOL3 LRRC29 B3GNT9 MIR328 FHOD1 FBXL8 PLEKHG4 ELMO3 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | KIAA0895L C16orf70 EXOC3L1 TMEM208 TRADD HSF4 E2F4 LOC105369155 NOL3 LRRC29 B3GNT9 MIR328 FHOD1 FBXL8 ELMO3 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | KIAA0895L EXOC3L1 MIR328 HSF4 E2F4 FBXL8 LOC105369155 NOL3 ELMO3 LRRC29 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | EXOC3L1 TMEM208 NUTF2 LOC105369155 NOL3 FHOD1 ENKD1 GFOD2 TSNAXIP1 EDC4 ELMO3 ATP6V0D1 KIAA0895L CARMIL2 FAM65A PARD6A LRRC29 LOC107984813 ACD MIR328 THAP11 SLC12A4 NRN1L FBXL8 LRRC36 AGRP LCAT LOC100131303 C16orf86 HSD11B2 DDX28 TRADD ZDHHC1 RANBP10 LOC100505942 DUS2 B3GNT9 DPEP2 CENPT CTCF PLEKHG4 DPEP3 C16orf70 PSKH1 HSF4 TPPP3 E2F4 SLC9A5 KCTD19 PSMB10 CTRL |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |