Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C12orf60 | NM_175874.3 | NP_787070.2 | ||
XM_005253322.4 | XP_005253379.1 | |||
XM_011520568.2 | XP_011518870.1 | |||
XM_011520569.2 | XP_011518871.1 | |||
XM_017018871.1 | XP_016874360.1 | |||
XM_017018872.1 | XP_016874361.1 | |||
XM_017018873.1 | XP_016874362.1 | |||
XM_017018874.1 | XP_016874363.1 | |||
AK056213.1 | ||||
AK289942.1 | ||||
BC015546.1 | AAH15546.1 | |||
BC029443.1 | AAH29443.1 | |||
BC038836.1 | AAH38836.1 |
Target Gene Details
Entrez Gene ID: | 440087 |
Gene Name: | single-pass membrane protein with coiled-coil domains 3 |
Gene Aliases: |
C12orf69 |
Location: |
Chr.12:14804650-14816761 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMCO3 | NM_001013698.2 | NP_001013720.2 | ||
XM_017019312.1 | XP_016874801.1 | |||
AK092637.1 | BAC03930.1 | |||
BC132752.1 | ||||
BC132754.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |