Assay Details
Target Gene Details
Entrez Gene ID: | 55738 |
Gene Name: | ADP ribosylation factor GTPase activating protein 1 |
Gene Aliases: |
ARF1GAP, HRIHFB2281 |
Location: |
Chr.20:63272785-63289793 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARFGAP1 | NM_001281482.1 | NP_001268411.1 | ||
NM_001281483.1 | NP_001268412.1 | |||
NM_001281484.1 | NP_001268413.1 | |||
NM_018209.3 | NP_060679.1 | |||
NM_175609.2 | NP_783202.1 | |||
XM_006723823.2 | XP_006723886.1 | |||
XM_011528901.1 | XP_011527203.1 | |||
XM_017027950.1 | XP_016883439.1 | |||
AK001629.1 | BAA91796.1 | |||
AK027268.1 | BAB55009.1 | |||
AK027441.1 | BAB55113.1 | |||
AK223427.1 | BAD97147.1 | |||
AK296351.1 | ||||
AK303454.1 | ||||
AK308161.1 | ||||
AY927573.1 | ||||
BC000786.1 | AAH00786.1 | |||
BC006085.1 | AAH06085.1 | |||
BC011876.1 | AAH11876.1 | |||
BC028233.1 | AAH28233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953306 | Chr.20:63151949 - 63571247 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv525279 | Chr.20:63221277 - 63323323 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv3446 | Chr.20:63237764 - 63282515 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv586564 | Chr.20:63276267 - 63321213 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv834032 | Chr.20:63213997 - 63308161 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)