Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | 1 | 71 | NP_001139528.1 |
NM_001146082.2 | 1 | 71 | NP_001139554.1 | |
NM_001146083.2 | 1 | 71 | NP_001139555.1 | |
NM_024298.4 | 1 | 71 | NP_077274.3 | |
XM_011527299.2 | 1 | 239 | XP_011525601.1 | |
XM_017027296.1 | 1 | 239 | XP_016882785.1 | |
AK055908.1 | BAB71043.1 | |||
DA338950.1 | 1 | 71 |
Target Gene Details
Entrez Gene ID: | 79042 |
Gene Name: | tRNA splicing endonuclease subunit 34 |
Gene Aliases: |
LENG5, PCH2C, SEN34, SEN34L |
Location: |
Chr.19:54189517-54194536 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSEN34 | XM_011527295.1 | XP_011525597.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066425 | Chr.19:54182441 - 54237431 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv2546 | Chr.19:54182614 - 54270671 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)