Assay Details
Target Gene Details
Entrez Gene ID: | 27151 |
Gene Name: | C3 and PZP like, alpha-2-macroglobulin domain containing 8 |
Gene Aliases: |
K-CAP, VIP |
Location: |
Chr.19:16892951-17026818 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPAMD8 | NM_015692.2 | 33 | 4501 | NP_056507.2 |
XM_011527917.1 | 33 | 4468 | XP_011526219.1 | |
XM_011527918.1 | 33 | 4504 | XP_011526220.1 | |
XM_011527919.1 | 33 | 4504 | XP_011526221.1 | |
XM_011527920.2 | 33 | 4504 | XP_011526222.1 | |
XM_011527921.2 | 33 | 4504 | XP_011526223.1 | |
XM_011527922.1 | 33 | 4504 | XP_011526224.1 | |
XM_011527923.1 | 33 | 4504 | XP_011526225.1 | |
XM_011527924.2 | 33 | 4504 | XP_011526226.1 | |
XM_011527925.1 | 17 | 2346 | XP_011526227.1 | |
XM_017026594.1 | 33 | 4504 | XP_016882083.1 | |
AB033109.1 | 33 | 4326 | BAA86597.1 | |
AY101765.1 | 33 | 4345 | AAM50084.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |