Assay Details
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM165 | NM_018475.4 | NP_060945.2 | ||
NR_073070.1 | ||||
XM_011534394.2 | XP_011532696.1 | |||
XM_017008412.1 | XP_016863901.1 | |||
AF132746.1 | AAL75947.1 | |||
AF183409.1 | AAG09678.1 | |||
AF220188.1 | AAF67653.1 | |||
AK026163.1 | ||||
AK054712.1 | ||||
AK290663.1 | ||||
AK295289.1 | ||||
BC003545.1 | AAH03545.2 | |||
BC104978.1 | AAI04979.1 | |||
BC104980.1 | AAI04981.1 | |||
BC107582.1 | AAI07583.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |