Assay Details
Target Gene Details
Entrez Gene ID: | 81025 |
Gene Name: | gap junction protein alpha 9 |
Gene Aliases: |
CX58, CX59, GJA10 |
Location: |
Chr.1:38874067-38881626 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GJA9 | NM_030772.4 | 2 | 248 | NP_110399.2 |
AF271261.1 | 1 | 211 | AAK55516.1 | |
AK312811.1 | 2 | 209 | ||
AL705384.1 | 2 | 248 | ||
BI463634.1 | 2 | 245 |
Target Gene Details
Entrez Gene ID: | 100527950 |
Gene Name: | GJA9-MYCBP readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:38862490-38881626 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GJA9-MYCBP | NR_037633.1 | 2 | 248 | |
NR_037634.1 | 2 | 248 | ||
NR_037635.1 | ||||
NR_037636.1 | ||||
NR_037637.1 | ||||
AK123051.1 | 2 | 242 | ||
FY210901.1 | ||||
FY211253.1 | ||||
FY211295.1 |
Target Gene Details
Entrez Gene ID: | 105378663 |
Gene Name: | uncharacterized LOC105378663 |
Gene Aliases: |
- |
Location: |
Chr.1:38873401-38876226 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105378663 | NR_135048.1 | 2 | 380 | |
BX115242.1 | 2 | 331 | ||
HY234287.1 | 1 | 85 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |