Assay Details
Target Gene Details
Entrez Gene ID: | 10850 |
Gene Name: | C-C motif chemokine ligand 27 |
Gene Aliases: |
ALP, CTACK, CTAK, ESKINE, ILC, PESKY, SCYA27 |
Location: |
Chr.9:34661883-34662692 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL27 | NM_006664.3 | NP_006655.1 | ||
AB010445.1 | BAA87046.1 | |||
AF082393.1 | AAD41238.1 | |||
AJ243542.1 | CAB46983.1 | |||
BC148263.1 | ||||
CA433929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain | DNAI1 SNORD121B LOC730098 ARID3C IL11RA UBAP2 C9orf24 UBAP1 CNTFR-AS1 KIAA1161 SIGMAR1 LOC114224 DCTN3 CCL21 CNTFR FAM205A SNORD121A UBE2R2 FAM219A DCAF12 CCL27 RPP25L NUDT2 CCL19 GALT KIF24 ENHO |
nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion | CNTFR-AS1 SIGMAR1 DCTN3 CCL27 CNTFR LOC730098 GALT RPP25L ARID3C IL11RA |
dgv12659n54 | Chr.9:34552439 - 34663141 on Build GRCh38 | Loss | CNTFR-AS1 SIGMAR1 DCTN3 CCL27 CNTFR GALT RPP25L ARID3C IL11RA |
nsv831544 | Chr.9:34650003 - 34818951 on Build GRCh38 | Gain | CCL19 PHF24 CCL27 CCL21 LOC730098 GALT FAM205A IL11RA |
nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain | CNTFR-AS1 SIGMAR1 DCTN3 CCL27 CNTFR LOC730098 GALT RPP25L ARID3C IL11RA |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |