Assay Details
Target Gene Details
Entrez Gene ID: | 2837 |
Gene Name: | urotensin 2 receptor |
Gene Aliases: |
GPR14, UR-2-R, UTR, UTR2 |
Location: |
Chr.17:82374277-82375586 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UTS2R | NM_018949.2 | 1 | 319 | NP_061822.1 |
BC136490.1 | 1 | 319 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059334 | Chr.17:82277007 - 82411683 on Build GRCh38 | Gain | UTS2R LOC105376790 OGFOD3 SECTM1 CD7 LOC101929511 TEX19 |
nsv833573 | Chr.17:82275053 - 82406183 on Build GRCh38 | Loss | UTS2R LOC105376790 OGFOD3 SECTM1 CD7 LOC101929511 TEX19 |
esv2760445 | Chr.17:82267669 - 82421456 on Build GRCh38 | Gain | UTS2R CSNK1D LOC105376790 HEXDC OGFOD3 SECTM1 CD7 LOC101929511 TEX19 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | C17orf62 PDE6G LOC105376790 FASN TSPAN10 LOC101929552 ARHGDIA TBCD UTS2R AATK ACTG1 SIRT7 ANAPC11 CD7 MRPL12 PCYT2 MAFG CCDC57 CSNK1D PYCR1 MIR6787 OGFOD3 SLC25A10 NARF BAHCC1 MYADML2 NPB P4HB WDR45B MCRIP1 C17orf89 CCDC137 SLC38A10 FN3KRP LINC00482 RAC3 LRRC45 TMEM105 FOXK2 GCGR DCXR AATK-AS1 FAAP100 HEXDC ASPSCR1 SECTM1 HGS MIR4525 ZNF750 NPLOC4 RAB40B LOC105371944 LOC105371925 MIR3186 MAFG-AS1 GPS1 MIR6786 RFNG STRA13 ARL16 ALYREF MIR4740 PPP1R27 CEP131 FSCN2 B3GNTL1 OXLD1 TEPSIN SLC16A3 LOC105376791 SNORD134 LOC100130370 TEX19 FN3K NOTUM DUS1L LOC101929511 |
esv3641423 | Chr.17:82369046 - 82398572 on Build GRCh38 | Gain | UTS2R LOC105376790 OGFOD3 |
nsv469561 | Chr.17:82251148 - 82472274 on Build GRCh38 | Loss | UTS2R CSNK1D C17orf62 LOC105376790 HEXDC OGFOD3 NARF SECTM1 CD7 LOC101929511 TEX19 |
nsv952394 | Chr.17:82297625 - 82408424 on Build GRCh38 | Deletion | UTS2R LOC105376790 OGFOD3 SECTM1 CD7 TEX19 |
nsv833572 | Chr.17:82204977 - 82388835 on Build GRCh38 | Loss | CCDC57 UTS2R CSNK1D LOC105376790 MIR6787 SECTM1 SLC16A3 CD7 LOC101929511 TEX19 |
nsv1067194 | Chr.17:82294731 - 82653129 on Build GRCh38 | Gain | UTS2R C17orf62 WDR45B LOC105376790 HEXDC OGFOD3 NARF SECTM1 FOXK2 CD7 TEX19 |
esv2762577 | Chr.17:82244836 - 82838431 on Build GRCh38 | Gain+Loss | CSNK1D C17orf62 LOC105376790 HEXDC OGFOD3 NARF SECTM1 LOC105376791 MIR4525 ZNF750 RAB40B LOC101929552 TEX19 FN3K TBCD UTS2R WDR45B FN3KRP FOXK2 CD7 LOC101929511 |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | C17orf62 LOC105376790 FASN LOC101929552 ARHGDIA TBCD UTS2R SIRT7 ANAPC11 CD7 PCYT2 MAFG CCDC57 CSNK1D PYCR1 MIR6787 OGFOD3 NARF MYADML2 NPB P4HB WDR45B MCRIP1 FN3KRP RAC3 LRRC45 FOXK2 DCXR HEXDC ASPSCR1 SECTM1 MIR4525 ZNF750 RAB40B LOC105371944 MAFG-AS1 GPS1 RFNG STRA13 ALYREF PPP1R27 B3GNTL1 SLC16A3 LOC105376791 SNORD134 TEX19 FN3K NOTUM DUS1L LOC101929511 METRNL |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |