Assay Details
Target Gene Details
Entrez Gene ID: | 558 |
Gene Name: | AXL receptor tyrosine kinase |
Gene Aliases: |
ARK, JTK11, Tyro7, UFO |
Location: |
Chr.19:41219199-41261767 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AXL | NM_001278599.1 | NP_001265528.1 | ||
NM_001699.5 | NP_001690.2 | |||
NM_021913.4 | NP_068713.2 | |||
AK309139.1 | ||||
AK310881.1 | ||||
BC032229.1 | AAH32229.1 | |||
M76125.1 | AAA61243.1 | |||
S65125.1 | AAB20305.1 | |||
X57019.1 | CAA40338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2014e59 | Chr.19:41229507 - 41231105 on Build GRCh38 | Duplication | AXL |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |