Assay Details
Target Gene Details
Entrez Gene ID: | 8372 |
Gene Name: | hyaluronoglucosaminidase 3 |
Gene Aliases: |
HYAL-3, LUCA-3, LUCA3 |
Location: |
Chr.3:50292828-50299468 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYAL3 | NM_001200029.1 | NP_001186958.1 | ||
NM_001200030.1 | NP_001186959.1 | |||
NM_001200031.1 | NP_001186960.1 | |||
NM_001200032.1 | NP_001186961.1 | |||
NM_003549.3 | NP_003540.2 | |||
AF036035.1 | AAD04257.1 | |||
AF040710.1 | AAC70915.1 | |||
BC005896.1 | AAH05896.1 | |||
BC012892.1 | AAH12892.1 | |||
DB319349.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759148 | Chr.3:50161045 - 50333946 on Build GRCh38 | Loss | LSMEM2 HYAL2 TUSC2 SEMA3B RASSF1 HYAL3 GNAT1 MIR566 MIR5787 SEMA3B-AS1 SEMA3F GNAI2 NAT6 IFRD2 SLC38A3 MIR6872 HYAL1 |
nsv523560 | Chr.3:50134964 - 50314769 on Build GRCh38 | Loss | LSMEM2 SEMA3B HYAL3 GNAT1 MIR566 MIR5787 SEMA3B-AS1 SEMA3F GNAI2 LOC107986035 SEMA3F-AS1 NAT6 IFRD2 SLC38A3 MIR6872 HYAL1 |
dgv8317n54 | Chr.3:50206077 - 50383123 on Build GRCh38 | Loss | RASSF1-AS1 LSMEM2 HYAL2 TUSC2 CYB561D2 ZMYND10 SEMA3B RASSF1 HYAL3 MIR5787 SEMA3B-AS1 GNAI2 NPRL2 NAT6 CACNA2D2 IFRD2 SLC38A3 MIR6872 TMEM115 HYAL1 |
nsv834693 | Chr.3:50251400 - 50470533 on Build GRCh38 | Loss | RASSF1-AS1 LSMEM2 HYAL2 TUSC2 CYB561D2 ZMYND10 SEMA3B RASSF1 HYAL3 SEMA3B-AS1 GNAI2 NPRL2 NAT6 CACNA2D2 IFRD2 MIR6872 TMEM115 HYAL1 |
nsv834692 | Chr.3:50161052 - 50356033 on Build GRCh38 | Loss | RASSF1-AS1 LSMEM2 HYAL2 TUSC2 CYB561D2 ZMYND10 SEMA3B RASSF1 HYAL3 GNAT1 MIR566 MIR5787 SEMA3B-AS1 SEMA3F GNAI2 NPRL2 NAT6 IFRD2 SLC38A3 MIR6872 TMEM115 HYAL1 |
dgv8316n54 | Chr.3:50115923 - 50299991 on Build GRCh38 | Loss | LSMEM2 RBM5 SEMA3B HYAL3 GNAT1 MIR566 MIR5787 SEMA3B-AS1 SEMA3F GNAI2 LOC107986035 SEMA3F-AS1 NAT6 IFRD2 SLC38A3 MIR6872 HYAL1 |
nsv470561 | Chr.3:50121341 - 50547568 on Build GRCh38 | Loss | LSMEM2 HYAL2 TUSC2 CYB561D2 ZMYND10 SEMA3B RASSF1 GNAT1 MIR566 MIR5787 SEMA3F GNAI2 NPRL2 LOC107986035 SLC38A3 RASSF1-AS1 HYAL3 SEMA3B-AS1 SEMA3F-AS1 NAT6 CACNA2D2 IFRD2 MIR6872 TMEM115 HYAL1 |
nsv523662 | Chr.3:50107518 - 50547568 on Build GRCh38 | Loss | LSMEM2 HYAL2 TUSC2 CYB561D2 RBM5 ZMYND10 SEMA3B RASSF1 GNAT1 MIR566 MIR5787 SEMA3F GNAI2 NPRL2 LOC107986035 SLC38A3 RASSF1-AS1 HYAL3 SEMA3B-AS1 SEMA3F-AS1 NAT6 CACNA2D2 IFRD2 MIR6872 TMEM115 HYAL1 |
nsv590255 | Chr.3:50210391 - 50399396 on Build GRCh38 | Loss | RASSF1-AS1 LSMEM2 HYAL2 TUSC2 CYB561D2 ZMYND10 SEMA3B RASSF1 HYAL3 MIR5787 SEMA3B-AS1 GNAI2 NPRL2 NAT6 CACNA2D2 IFRD2 SLC38A3 MIR6872 TMEM115 HYAL1 |
nsv954855 | Chr.3:50152468 - 50439169 on Build GRCh38 | Deletion | LSMEM2 HYAL2 TUSC2 CYB561D2 ZMYND10 SEMA3B RASSF1 GNAT1 MIR566 MIR5787 SEMA3F GNAI2 NPRL2 LOC107986035 SLC38A3 RASSF1-AS1 HYAL3 SEMA3B-AS1 SEMA3F-AS1 NAT6 CACNA2D2 IFRD2 MIR6872 TMEM115 HYAL1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |