Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMDHD2 | NM_001145815.1 | 2 | 242 | NP_001139287.1 |
NM_015944.3 | 2 | 242 | NP_057028.2 | |
XM_017023262.1 | 2 | 253 | XP_016878751.1 | |
XM_017023263.1 | 2 | 253 | XP_016878752.1 | |
XM_017023264.1 | 2 | 253 | XP_016878753.1 | |
XM_017023265.1 | 2 | 253 | XP_016878754.1 | |
XM_017023266.1 | 2 | 201 | XP_016878755.1 | |
XM_017023267.1 | XP_016878756.1 | |||
AF132948.1 | 2 | 235 | AAD27723.1 | |
AK296877.1 | 2 | 214 | ||
AK300747.1 | ||||
BC018734.1 | 2 | 178 | AAH18734.1 | |
CN362870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | HBQ1 C1QTNF8 WFIKKN1 MSLN NTHL1 LOC105371047 PRSS29P NUBP2 RPUSD1 CCDC154 LUC7L WDR90 IFT140 LOC107984876 RHOT2 RAB11FIP3 HBZ PKD1 NPW MSRB1 RAB40C MIR662 FLJ42627 MIR6767 SNORA10 MIR3180-5 RPL3L LOC107987233 IGFALS MIR4717 DDX11L10 MRPL28 SPSB3 DNASE1L2 AMDHD2 LMF1-AS1 HBA2 LOC105371045 MEIOB TSR3 AXIN1 JMJD8 SLC9A3R2 SNORA78 TSC2 PDIA2 LINC00254 DECR2 LOC100134368 LOC652276 PTX4 ECI1 CHTF18 TMEM8A HN1L SSTR5 SYNGR3 MIR3176 HBM MCRIP2 SNORA64 PIGQ SNORD60 TMEM204 PGP SNHG19 RGS11 SNRNP25 NPRL3 TBC1D24 CACNA1H EME2 FAHD1 MIR1225 LOC105371046 TPSD1 ARHGDIG WASIR2 STUB1 GNG13 ATP6V0C METRN FAM234A CLCN7 MIR6768 LOC106660606 MLST8 RHBDL1 MIR6859-4 CCDC78 NME3 HAGH PRR25 PRSS27 NDUFB10 ERVK13-1 RAB26 SOX8 LOC105371038 MAPK8IP3 C16orf91 MIR3177 NME4 LMF1 LOC100287175 FBXL16 RPS2 RPL23AP5 LINC00235 PRR35 MIR4516 FAM173A POLR3K UNKL LOC101929280 MIR940 SNHG9 RHBDF1 BRICD5 C16orf59 LOC105371049 MIR6511B1 TBL3 GFER ZNF598 MIR5587 NARFL CEMP1 MPG WDR24 TELO2 TRAF7 HBA1 CAPN15 MIR3677 HS3ST6 ABCA17P NOXO1 CCNF METTL26 HAGHL LOC729652 CRAMP1 TPSB2 LOC105371184 KCTD5 MRPS34 UBE2I SSTR5-AS1 ABCA3 TPSAB1 MIR3178 RNPS1 GNPTG NTN3 PDPK1 BAIAP3 RNF151 TPSG1 CASKIN1 E4F1 NHLRC4 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | FLJ42627 PRSS41 ATP6V0C LOC652276 CEMP1 KCTD5 TCEB2 AMDHD2 PRSS21 TBC1D24 MIR3178 PRSS33 PRSS27 ERVK13-1 PDPK1 SRRM2 SRRM2-AS1 |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | FLJ42627 TBC1D24 MIR3178 ATP6V0C ERVK13-1 PDPK1 LOC652276 CEMP1 AMDHD2 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | TSC2 BRICD5 ATP6V0C C16orf59 LOC105371049 MIR6511B1 MIR6768 LOC106660606 LOC652276 CEMP1 ECI1 MLST8 TRAF7 MIR3677 RAB26 ABCA17P PKD1 CCNF LOC729652 SNORD60 MIR6767 MIR3180-5 PGP MIR4717 SNHG19 DNASE1L2 AMDHD2 ABCA3 TBC1D24 MIR3178 MIR4516 RNPS1 MIR1225 NTN3 PDPK1 MIR940 CASKIN1 E4F1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |