Assay Details
Target Gene Details
Entrez Gene ID: | 5734 |
Gene Name: | prostaglandin E receptor 4 |
Gene Aliases: |
EP4, EP4R |
Location: |
Chr.5:40679930-40740932 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGER4 | NM_000958.2 | 2 | 1022 | NP_000949.1 |
XM_017009656.1 | 1 | 473 | XP_016865145.1 | |
XM_017009657.1 | 1 | 473 | XP_016865146.1 | |
XM_017009658.1 | 1 | 473 | XP_016865147.1 | |
XM_017009659.1 | 1 | 473 | XP_016865148.1 | |
AY429109.1 | 1 | 430 | AAR07904.1 | |
BC027934.1 | 2 | 1022 | ||
BC101534.1 | 2 | 489 | AAI01535.1 | |
BC113523.1 | 2 | 489 | ||
D28472.1 | 2 | 514 | BAA05834.1 | |
L25124.1 | 2 | 1015 | AAA36438.1 | |
L28175.1 | 2 | 818 | AAA36434.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |