Assay Details
Target Gene Details
Entrez Gene ID: | 8507 |
Gene Name: | ectodermal-neural cortex 1 |
Gene Aliases: |
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10 |
Location: |
Chr.5:74627406-74641424 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENC1 | NM_001256574.1 | 3 | 2914 | NP_001243503.1 |
NM_001256575.1 | 3 | 2902 | NP_001243504.1 | |
NM_001256576.1 | 2 | 2641 | NP_001243505.1 | |
NM_003633.3 | 2 | 2777 | NP_003624.1 | |
NR_046318.1 | ||||
XM_011543696.2 | 3 | 2985 | XP_011541998.1 | |
XM_011543697.2 | 3 | 2831 | XP_011541999.1 | |
AF005381.1 | 3 | 1802 | AAC64498.1 | |
AF010314.1 | 2 | 2070 | AAC39532.1 | |
AF059611.1 | 2 | 2045 | AAC26109.1 | |
AK096578.1 | 2 | 2777 | ||
AK295128.1 | 2 | 1946 | ||
AK299624.1 | 2 | 971 | ||
AK312608.1 | 2 | 2079 | ||
AY049781.1 | 1 | 1650 | AAL15438.1 | |
BC000418.2 | 2 | 2047 | AAH00418.1 | |
BT007392.1 | 1 | 1646 | AAP36056.1 | |
DA785554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830351 | Chr.5:74485729 - 74637007 on Build GRCh38 | Loss | LINC01331 ENC1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |