Assay Details
Target Gene Details
Entrez Gene ID: | 1010 |
Gene Name: | cadherin 12 |
Gene Aliases: |
CDHB |
Location: |
Chr.5:21750191-22853622 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH12 | NM_001317227.1 | 12 | 2806 | NP_001304156.1 |
NM_001317228.1 | 12 | 2801 | NP_001304157.1 | |
NM_004061.4 | 15 | 3435 | NP_004052.2 | |
XM_011513927.2 | 13 | 2712 | XP_011512229.1 | |
XM_017008920.1 | 14 | 3062 | XP_016864409.1 | |
XM_017008921.1 | 15 | 3155 | XP_016864410.1 | |
AK092176.1 | 10 | 2085 | ||
AK295123.1 | 12 | 2801 | ||
AK314800.1 | ||||
BC047608.1 | 15 | 3140 | AAH47608.1 | |
L33477.1 | 15 | 3436 | AAB48539.1 | |
L34057.1 | 11 | 2441 | AAA35623.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |