Assay Details
Target Gene Details
Entrez Gene ID: | 53942 |
Gene Name: | contactin 5 |
Gene Aliases: |
HNB-2s, NB-2 |
Location: |
Chr.11:99020904-100358885 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNTN5 | NM_001243270.1 | 1 | 258 | NP_001230199.1 |
NM_001243271.1 | 1 | 258 | NP_001230200.1 | |
NM_014361.3 | 1 | 258 | NP_055176.1 | |
XM_011542871.1 | 1 | 258 | XP_011541173.1 | |
XM_011542873.1 | 1 | 258 | XP_011541175.1 | |
XM_017017926.1 | 1 | 330 | XP_016873415.1 | |
XM_017017927.1 | 1 | 330 | XP_016873416.1 | |
XM_017017928.1 | 1 | 330 | XP_016873417.1 | |
XM_017017929.1 | 1 | 258 | XP_016873418.1 | |
AK308788.1 | 1 | 258 | ||
BC039255.1 | 1 | 93 | AAH39255.1 | |
BC130619.1 | ||||
BC144185.1 | ||||
BC144186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428265 | Chr.11:98936705 - 99229707 on Build GRCh38 | Loss | CNTN5 |
esv3580039 | Chr.11:98983622 - 99023292 on Build GRCh38 | Gain | CNTN5 |
nsv832242 | Chr.11:98893493 - 99087654 on Build GRCh38 | Loss | CNTN5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |