Assay Details
Target Gene Details
Entrez Gene ID: | 8412 |
Gene Name: | breast cancer anti-estrogen resistance 3 |
Gene Aliases: |
NSP2, SH2D3B |
Location: |
Chr.1:93561741-93847150 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAR3 | NM_001261408.1 | 9 | 1574 | NP_001248337.1 |
NM_001261409.1 | 7 | 1560 | NP_001248338.1 | |
NM_001261410.1 | 5 | 1227 | NP_001248339.1 | |
NM_001308251.1 | 3 | 547 | NP_001295180.1 | |
NM_003567.3 | 7 | 1537 | NP_003558.1 | |
XM_011542251.2 | 6 | 1176 | XP_011540553.1 | |
XM_017002480.1 | 9 | 1950 | XP_016857969.1 | |
AF124250.1 | 7 | 1529 | AAD28245.1 | |
AK027785.1 | 3 | 795 | ||
AK055442.1 | 7 | 1560 | ||
AK301570.1 | 3 | 520 | ||
AK310970.1 | 3 | 547 | ||
AL833121.1 | 5 | 1180 | ||
AY358996.1 | 3 | 467 | AAQ89355.1 | |
BC039895.1 | 7 | 1430 | AAH39895.1 | |
DA745974.1 | 3 | 510 | ||
U92715.1 | 7 | 1345 | AAC39777.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |