Assay Details
Target Gene Details
Entrez Gene ID: | 161931 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
TENRL |
Location: |
Chr.16:84191117-84197166 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAD2 | NM_001145400.1 | NP_001138872.1 | ||
NM_139174.3 | NP_631913.3 | |||
XM_005255814.1 | XP_005255871.1 | |||
XM_011522919.1 | XP_011521221.1 | |||
XM_011522920.1 | XP_011521222.1 | |||
AF447586.1 | AAM22869.1 | |||
AK093046.1 | BAC04032.1 | |||
AK093523.1 | 1 | 174 | ||
AK131102.1 | 2 | 2972 | BAC85152.1 | |
AK315171.1 | ||||
BC033491.1 | AAH33491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639413 | Chr.16:84172318 - 84253359 on Build GRCh38 | Loss | DNAAF1 ADAD2 KCNG4 TAF1C |
nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss | HSDL1 DNAAF1 ADAD2 KCNG4 LOC729887 MBTPS1 TAF1C |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | HSDL1 DNAAF1 SLC38A8 ADAD2 LOC105371371 KCNG4 NECAB2 OSGIN1 LOC729887 MBTPS1 MLYCD TAF1C |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | LOC105376777 HSDL1 DNAAF1 SLC38A8 ADAD2 KCNG4 WFDC1 LOC729887 MBTPS1 TAF1C |
nsv523437 | Chr.16:84176132 - 84196770 on Build GRCh38 | Loss | DNAAF1 ADAD2 TAF1C |
esv3639414 | Chr.16:84176251 - 84352273 on Build GRCh38 | Loss | LOC105376777 DNAAF1 ADAD2 KCNG4 WFDC1 TAF1C |
nsv457597 | Chr.16:84182086 - 84215151 on Build GRCh38 | Loss | ADAD2 TAF1C |
esv3639412 | Chr.16:84170074 - 84221406 on Build GRCh38 | Loss | DNAAF1 ADAD2 KCNG4 TAF1C |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | COX4I1 MIR1910 FOXL1 FAM92B LOC101928474 COTL1 WFDC1 LOC105371374 TAF1C ZDHHC7 GINS2 IRF8 LINC01082 KCNG4 LINC00917 MIR5093 FOXC2-AS1 LOC101928557 LOC101928614 USP10 CRISPLD2 LOC105376777 C16orf74 EMC8 FOXF1 LOC102724344 FLJ30679 LINC00311 DNAAF1 LINC01081 TLDC1 ADAD2 LOC440390 MTHFSD FENDRR KIAA0513 ATP2C2 LOC400548 KLHL36 FOXC2 LOC146513 GSE1 MIR6774 |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | HSDL1 DNAAF1 SLC38A8 ADAD2 KCNG4 LOC729887 MBTPS1 TAF1C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |