Assay Details
Target Gene Details
Entrez Gene ID: | 64787 |
Gene Name: | EPS8 like 2 |
Gene Aliases: |
EPS8R2 |
Location: |
Chr.11:705217-727727 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS8L2 | NM_022772.3 | 10 | 1071 | NP_073609.2 |
XM_017018130.1 | 11 | 1288 | XP_016873619.1 | |
XM_017018131.1 | 10 | 1159 | XP_016873620.1 | |
XM_017018132.1 | 11 | 1129 | XP_016873621.1 | |
AK025588.1 | 10 | 989 | BAB15180.1 | |
AK025824.1 | 10 | 989 | BAB15248.1 | |
AK094539.1 | 3 | 256 | ||
AK122903.1 | 7 | 1267 | ||
AK222903.1 | 10 | 984 | BAD96623.1 | |
AK225311.1 | 10 | 989 | ||
AK294041.1 | 10 | 1036 | ||
AY074929.1 | 10 | 1096 | AAL76118.1 | |
BC080636.1 | 10 | 1075 | AAH80636.1 | |
BC093878.1 | 9 | 864 | AAH93878.1 | |
BC101481.1 | 9 | 864 | AAI01482.1 | |
BC143242.1 | 9 | 912 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522733 | Chr.11:702250 - 752059 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832043 | Chr.11:631027 - 758602 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv552869 | Chr.11:702097 - 727446 on Build GRCh38 | Loss |
![]() ![]() |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)