Assay Details
Target Gene Details
Entrez Gene ID: | 64788 |
Gene Name: | lipase maturation factor 1 |
Gene Aliases: |
C16orf26, HMFN1876, JFP11, TMEM112, TMEM112A |
Location: |
Chr.16:853632-981615 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LMF1 | NM_022773.2 | 11 | 2130 | NP_073610.2 |
NR_033645.1 | 12 | 2259 | ||
NR_033646.1 | 13 | 2437 | ||
XM_006720928.2 | 11 | 2403 | XP_006720991.1 | |
XM_006720929.3 | 12 | 2284 | XP_006720992.1 | |
XM_011522617.2 | 9 | 1779 | XP_011520919.1 | |
XM_017023576.1 | 10 | 1894 | XP_016879065.1 | |
XM_017023577.1 | 12 | 2559 | XP_016879066.1 | |
XM_017023578.1 | 10 | 1979 | XP_016879067.1 | |
XM_017023579.1 | 10 | 1987 | XP_016879068.1 | |
XM_017023580.1 | 13 | 2709 | XP_016879069.1 | |
AK022743.1 | 11 | 2128 | BAB14218.1 | |
AK093050.1 | 8 | 1637 | ||
AK125201.1 | 12 | 2261 | ||
CA427159.1 | 1 | 491 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571046 | Chr.16:828324 - 1202441 on Build GRCh38 | Gain | SOX8 SSTR5-AS1 CACNA1H LMF1 LMF1-AS1 LOC107984876 SSTR5 C1QTNF8 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR6859-4 MIR5587 TPSD1 DNASE1L2 NME3 FAHD1 NHLRC4 TELO2 CCDC154 RGS11 SNORA64 LOC105371045 NTN3 SNORD60 MLST8 LOC101929280 HBA2 LOC100134368 LOC105371047 NDUFB10 HBA1 MAPK8IP3 PIGQ ABCA17P UNKL MIR662 POLR3K LOC105371049 SNORA10 TBC1D24 IGFALS RHOT2 UBE2I SSTR5 GNPTG CRAMP1 JMJD8 PGP TMEM8A KCTD5 SNHG19 FBXL16 MIR6768 NPW SYNGR3 TRAF7 MIR6511B1 MIR4717 RAB26 MCRIP2 CCDC78 NPRL3 HBM HAGHL GFER WASIR2 DECR2 LOC107984876 METTL26 LOC729652 PKD1 WFIKKN1 TPSG1 ERVK13-1 FLJ42627 FAM173A MIR6767 ABCA3 WDR24 PDPK1 C16orf59 TPSAB1 MIR940 RHBDF1 MIR3176 C1QTNF8 MIR3178 LOC652276 BAIAP3 MSRB1 LUC7L CAPN15 PRSS27 ECI1 HN1L MRPL28 MRPS34 RHBDL1 WDR90 CCNF C16orf91 RNPS1 RPUSD1 LOC100287175 HBQ1 CACNA1H FAM234A TMEM204 LMF1 CHTF18 GNG13 LINC00254 NARFL ZNF598 RAB11FIP3 RPL3L RNF151 NTHL1 SOX8 MIR3677 SNRNP25 HAGH ATP6V0C RPS2 SLC9A3R2 PRR25 LMF1-AS1 ARHGDIG PDIA2 NOXO1 NUBP2 IFT140 TPSB2 MSLN LOC105371184 HBZ LOC106660606 NME4 LOC105371038 TSC2 PRR35 METRN EME2 MIR3177 CEMP1 MIR4516 SPSB3 CLCN7 DDX11L10 MIR3180-5 RPL23AP5 BRICD5 AMDHD2 CASKIN1 SNHG9 SSTR5-AS1 PRSS29P LOC105371046 MPG MIR1225 STUB1 HS3ST6 LOC107987233 LINC00235 AXIN1 RAB40C TSR3 TBL3 SNORA78 PTX4 MEIOB E4F1 |
esv3637586 | Chr.16:798995 - 892120 on Build GRCh38 | Loss | LMF1 GNG13 PRR25 |
nsv1704 | Chr.16:836423 - 867931 on Build GRCh38 | Insertion | LMF1 |
nsv1046349 | Chr.16:819378 - 942590 on Build GRCh38 | Loss | LMF1 LMF1-AS1 LOC107984876 |
nsv952898 | Chr.16:495001 - 1132900 on Build GRCh38 | Deletion | LOC105371184 FBXL16 LOC105371038 MIR5587 PRR35 METRN CAPN15 NHLRC4 MCRIP2 CCDC78 RHBDL1 WDR90 HAGHL LOC101929280 LOC107984876 RPUSD1 METTL26 LOC100287175 SSTR5-AS1 PIGQ LMF1 MIR662 WFIKKN1 CHTF18 GNG13 NARFL STUB1 RHOT2 RAB11FIP3 SSTR5 LOC107987233 FAM173A LINC00235 SOX8 RAB40C WDR24 PRR25 JMJD8 LMF1-AS1 MIR3176 C1QTNF8 MSLN |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |