Assay Details
Target Gene Details
Entrez Gene ID: | 161931 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
TENRL |
Location: |
Chr.16:84191117-84197166 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAD2 | NM_001145400.1 | NP_001138872.1 | ||
NM_139174.3 | NP_631913.3 | |||
XM_005255814.1 | XP_005255871.1 | |||
XM_011522919.1 | XP_011521221.1 | |||
XM_011522920.1 | XP_011521222.1 | |||
AF447586.1 | AAM22869.1 | |||
AK093046.1 | BAC04032.1 | |||
AK093523.1 | ||||
AK131102.1 | BAC85152.1 | |||
AK315171.1 | ||||
BC033491.1 | AAH33491.1 | |||
BU731201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639413 | Chr.16:84172318 - 84253359 on Build GRCh38 | Loss | KCNG4 TAF1C DNAAF1 ADAD2 |
nsv518342 | Chr.16:84194593 - 84196629 on Build GRCh38 | Loss | ADAD2 |
nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss | MBTPS1 LOC729887 KCNG4 TAF1C HSDL1 DNAAF1 ADAD2 |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | MBTPS1 NECAB2 MLYCD LOC105371371 LOC729887 KCNG4 TAF1C SLC38A8 HSDL1 DNAAF1 ADAD2 OSGIN1 |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | WFDC1 MBTPS1 LOC729887 KCNG4 TAF1C LOC105376777 SLC38A8 HSDL1 DNAAF1 ADAD2 |
nsv523437 | Chr.16:84176132 - 84196770 on Build GRCh38 | Loss | TAF1C DNAAF1 ADAD2 |
esv3639414 | Chr.16:84176251 - 84352273 on Build GRCh38 | Loss | WFDC1 KCNG4 TAF1C LOC105376777 DNAAF1 ADAD2 |
nsv457597 | Chr.16:84182086 - 84215151 on Build GRCh38 | Loss | TAF1C ADAD2 |
esv3639412 | Chr.16:84170074 - 84221406 on Build GRCh38 | Loss | KCNG4 TAF1C DNAAF1 ADAD2 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | LINC00917 KLHL36 ZDHHC7 TAF1C LOC101928474 LINC00311 LOC400548 LINC01081 LINC01082 USP10 FLJ30679 FENDRR MIR1910 CRISPLD2 LOC105376777 LOC105371374 COX4I1 C16orf74 LOC146513 COTL1 EMC8 LOC101928614 WFDC1 ATP2C2 FOXC2 MIR6774 FAM92B IRF8 FOXF1 TLDC1 GINS2 KIAA0513 LOC440390 ADAD2 LOC102724344 LOC101928557 KCNG4 GSE1 FOXC2-AS1 FOXL1 DNAAF1 MIR5093 MTHFSD |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | MBTPS1 LOC729887 KCNG4 TAF1C SLC38A8 HSDL1 DNAAF1 ADAD2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |