Assay Details
Target Gene Details
Entrez Gene ID: | 27151 |
Gene Name: | C3 and PZP like, alpha-2-macroglobulin domain containing 8 |
Gene Aliases: |
K-CAP, VIP |
Location: |
Chr.19:16892951-17026818 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPAMD8 | NM_015692.2 | 42 | 5765 | NP_056507.2 |
XM_011527917.1 | 42 | 5732 | XP_011526219.1 | |
XM_011527918.1 | 42 | 6688 | XP_011526220.1 | |
XM_011527919.1 | 42 | 6866 | XP_011526221.1 | |
XM_011527920.2 | 42 | 8040 | XP_011526222.1 | |
XM_011527924.2 | 42 | 7966 | XP_011526226.1 | |
XM_011527925.1 | 26 | 3610 | XP_011526227.1 | |
AB033109.1 | 42 | 5590 | BAA86597.1 | |
AI670110.1 | 1 | 232 | ||
AK075099.1 | 9 | 2258 | BAC11400.1 | |
AK092632.1 | 3 | 1824 | ||
AK124052.1 | 2 | 3266 | ||
AY101765.1 | 42 | 5609 | AAM50084.1 | |
CA421477.1 | 1 | 235 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |