Assay Details
Target Gene Details
Entrez Gene ID: | 26019 |
Gene Name: | UPF2 regulator of nonsense transcripts homolog (yeast) |
Gene Aliases: |
HUPF2, RENT2, smg-3 |
Location: |
Chr.10:11920022-12043170 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UPF2 | NM_015542.3 | 22 | 4017 | NP_056357.1 |
NM_080599.2 | 22 | 4187 | NP_542166.1 | |
XM_017016034.1 | 21 | 3673 | XP_016871523.1 | |
AB037829.1 | 21 | 4386 | ||
AF301013.1 | 22 | 4010 | AAG33225.1 | |
AF318574.1 | 22 | 3987 | AAG60689.1 | |
AK000764.1 | 7 | 848 | BAA91369.1 | |
AK074375.1 | 7 | 782 | BAB85064.1 | |
AK096191.1 | BAC04721.1 | |||
AK124930.1 | 1 | 1075 | ||
AK295209.1 | 9 | 1086 | ||
AL080198.1 | 16 | 2187 | CAB45771.1 | |
BC114964.1 | 22 | 3981 | ||
BC115737.1 | 22 | 3981 | ||
BX647325.1 | 1 | 1000 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |