Assay Details
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001170695.1 | 9 | 1117 | NP_001164166.1 |
NM_001170696.1 | 8 | 988 | NP_001164167.1 | |
NM_001170697.1 | 9 | 1123 | NP_001164168.1 | |
NM_001170698.1 | 9 | 1138 | NP_001164169.1 | |
NM_001170699.1 | 8 | 1259 | NP_001164170.1 | |
NM_001321336.1 | 9 | 1407 | NP_001308265.1 | |
NM_001321337.1 | 9 | 1455 | NP_001308266.1 | |
NM_032598.4 | 9 | 1179 | NP_115987.2 | |
AF367472.1 | 9 | 1071 | AAK53408.1 | |
AK057485.1 | 1 | 1547 | ||
AK295299.1 | 9 | 1407 | ||
AK301892.1 | ||||
AY032684.1 | 8 | 937 | AAK51120.1 | |
AY035867.1 | 9 | 1120 | AAK61373.1 | |
AY035868.1 | 9 | 1179 | AAK61374.1 | |
BC029483.1 | 9 | 1064 | AAH29483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss | OR1E2 OR1A1 OR1A2 OR3A4P OR1D4 TRPV3 ASPA OR3A1 OR1E1 OR3A3 TRPV1 SPATA22 OR3A2 |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain | CCDC92B OR1E2 TAX1BP3 RAP1GAP2 P2RX5-TAX1BP3 P2RX5 MIR1253 OR1D2 MIR6776 OR1D4 EMC6 ITGAE ASPA OR3A1 LOC105371592 OR1E1 TRPV1 SPATA22 OR1G1 ATP2A3 CAMKK1 PAFAH1B1 SHPK OR1A1 LOC105371591 OR1A2 OR1D5 OR3A4P LOC101927911 TRPV3 GSG2 OR3A3 CLUH OR3A2 NCBP3 P2RX1 CTNS |
nsv574232 | Chr.17:3254715 - 3503170 on Build GRCh38 | Gain | OR1E2 ASPA OR3A1 OR1E1 OR3A3 SPATA22 OR3A4P OR3A2 |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion | CCDC92B OR1E2 PAFAH1B1 OR1A1 LOC105371591 RAP1GAP2 OR1A2 MIR1253 OR1D5 OR3A4P OR1D2 MIR6776 LOC101927911 OR1D4 TRPV3 ASPA OR3A1 LOC105371592 OR1E1 OR3A3 SPATA22 OR1G1 CLUH OR3A2 |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain | OR1E2 TAX1BP3 SHPK P2RX5-TAX1BP3 P2RX5 TRPV3 EMC6 ITGAE ASPA GSG2 OR1E1 OR3A3 TRPV1 SPATA22 CAMKK1 OR3A2 NCBP3 CTNS |
nsv521277 | Chr.17:3390044 - 3468885 on Build GRCh38 | Gain | OR1E2 OR1E1 OR3A3 SPATA22 |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss | OR1E2 ASPA OR1E1 OR3A3 SPATA22 OR3A2 TRPV3 |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss | OR1E2 ASPA SHPK OR3A3 TRPV1 SPATA22 TRPV3 |
nsv1059413 | Chr.17:3365789 - 3453922 on Build GRCh38 | Loss | OR1E2 OR1E1 OR3A3 SPATA22 OR3A2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |