Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC1 | NM_001130107.1 | NP_001123579.1 | ||
NM_182923.3 | NP_891553.2 | |||
AB451299.1 | ||||
AB451431.1 | ||||
AY180165.3 | AAO62550.1 | |||
AY180168.1 | AAO62553.1 | |||
BM988840.1 | ||||
L04733.1 | AAA16576.1 |
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | ||||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | LOC107984648 XRCC3 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | LOC107984648 XRCC3 ZFYVE21 KLC1 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | LOC107984648 XRCC3 ZFYVE21 KLC1 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | LOC107984648 XRCC3 ZFYVE21 KLC1 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | MARK3 RD3L MIR4710 LOC105370708 SNORA28 C14orf79 AKT1 LOC101929634 MIR6765 AHNAK2 LOC107984648 KIF26A LINC00637 MIR203B C14orf2 SIVA1 PLD4 ZFYVE21 NUDT14 CKB BTBD6 EIF5 LOC105370687 C14orf180 TDRD9 TMEM179 PPP1R13B BAG5 XRCC3 GPR132 ADSSL1 PACS2 LINC00638 ZBTB42 INF2 LOC102723354 CEP170B TRMT61A MIR203A ASPG JAG2 LOC105378183 LOC388022 APOPT1 KLC1 LOC105370691 CDCA4 BRF1 |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | LOC107984648 LINC00637 RD3L TDRD9 C14orf2 PPP1R13B XRCC3 ZFYVE21 KLC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |