Assay Details
Target Gene Details
Entrez Gene ID: | 1649 |
Gene Name: | DNA damage inducible transcript 3 |
Gene Aliases: |
CEBPZ, CHOP, CHOP-10, CHOP10, GADD153 |
Location: |
Chr.12:57516588-57520517 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDIT3 | NM_001195053.1 | NP_001181982.1 | ||
NM_001195054.1 | NP_001181983.1 | |||
NM_001195055.1 | NP_001181984.1 | |||
NM_001195056.1 | NP_001181985.1 | |||
NM_001195057.1 | NP_001181986.1 | |||
NM_004083.5 | NP_004074.2 | |||
AA054097.1 | ||||
AA476561.1 | ||||
AK316581.1 | ||||
AV729744.1 | ||||
AY461580.1 | ||||
BC003637.2 | AAH03637.1 | |||
BM464290.1 | ||||
BU664077.1 | ||||
CN482428.1 | ||||
S40706.1 | AAB22646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv559036 | Chr.12:57517429 - 57522259 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)