Assay Details
Target Gene Details
Entrez Gene ID: | 1201 |
Gene Name: | CLN3, battenin |
Gene Aliases: |
BTS, JNCL |
Location: |
Chr.16:28466653-28492302 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | NUPR1 APOBR CLN3 SULT1A1 SGF29 SULT1A2 IL27 |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | XPO6 SPNS1 RRN3P2 MIR6862-1 EIF3CL ATXN2L ATP2A1-AS1 SH2B1 MIR6862-2 NFATC2IP LOC101928215 NPIP MIR4517 NPIPB8 SGF29 SULT1A2 IL27 NUPR1 APOBR EIF3C MIR4721 SULT1A1 NPIPB6 CD19 ATP2A1 SBK1 TUFM CLN3 NPIPB9 LAT RABEP2 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | SPNS1 RRN3P2 MIR6862-1 EIF3CL ATXN2L ATP2A1-AS1 SH2B1 MIR6862-2 NFATC2IP LOC101928215 NPIP MIR4517 NPIPB8 SGF29 SULT1A2 IL27 NUPR1 APOBR EIF3C MIR4721 SULT1A1 CD19 ATP2A1 TUFM CLN3 NPIPB9 LOC107984832 LAT RABEP2 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | NUPR1 APOBR CLN3 MIR6862-1 EIF3CL NPIPB6 SGF29 IL27 |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | APOBR CLN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |