Assay Details
Target Gene Details
Entrez Gene ID: | 7306 |
Gene Name: | tyrosinase related protein 1 |
Gene Aliases: |
CAS2, CATB, GP75, OCA3, TRP, TRP1, TYRP, b-PROTEIN |
Location: |
Chr.9:12693385-12710266 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TYRP1 | NM_000550.2 | 2 | 138 | NP_000541.1 |
BC052608.1 | 1 | 57 | AAH52608.1 | |
CR407683.1 | CAG28611.1 | |||
DA278582.1 | 2 | 138 | ||
X51420.1 | 2 | 89 | CAA35785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891612 | Chr.9:12213879 - 13226946 on Build GRCh38 | Gain | SNORD137 LURAP1L-AS1 MPDZ LURAP1L TYRP1 |
dgv12568n54 | Chr.9:12628213 - 12712157 on Build GRCh38 | Loss | LURAP1L-AS1 TYRP1 |
nsv1021844 | Chr.9:12356017 - 13444976 on Build GRCh38 | Gain | SNORD137 LINC01235 LURAP1L-AS1 MPDZ LURAP1L TYRP1 |
nsv1016570 | Chr.9:12576095 - 12722388 on Build GRCh38 | Loss | LURAP1L-AS1 TYRP1 |
nsv831513 | Chr.9:12531673 - 12695747 on Build GRCh38 | Gain | TYRP1 |
esv3619718 | Chr.9:12621841 - 12766938 on Build GRCh38 | Gain | LURAP1L-AS1 TYRP1 |
esv2422212 | Chr.9:12448789 - 13127739 on Build GRCh38 | Duplication | SNORD137 LURAP1L-AS1 MPDZ LURAP1L TYRP1 |
esv2759671 | Chr.9:12235048 - 12784093 on Build GRCh38 | Loss | LURAP1L-AS1 LURAP1L TYRP1 |
nsv613613 | Chr.9:12641987 - 12773264 on Build GRCh38 | Loss | LURAP1L-AS1 TYRP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |