Assay Details
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | 5 | 633 | NP_001093588.1 |
NM_001100119.1 | 6 | 727 | NP_001093589.1 | |
NM_005432.3 | 6 | 690 | NP_005423.1 | |
XM_005268046.2 | 5 | 727 | XP_005268103.1 | |
XM_011537138.2 | 6 | 673 | XP_011535440.1 | |
AF035586.1 | 5 | 618 | AAC05368.1 | |
AK022829.1 | 6 | 619 | ||
AK023646.1 | 6 | 663 | ||
AK124498.1 | 4 | 1107 | ||
AK126706.1 | 1 | 1759 | ||
BC001036.1 | 6 | 662 | AAH01036.1 | |
BC011725.2 | 5 | 593 | AAH11725.1 | |
BT007417.1 | 3 | 310 | AAP36085.1 | |
BX161398.1 | 5 | 602 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | XRCC3 LOC107984648 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | XRCC3 LOC107984648 KLC1 ZFYVE21 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | XRCC3 LOC107984648 KLC1 ZFYVE21 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | XRCC3 LOC107984648 KLC1 ZFYVE21 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | SNORA28 RD3L C14orf2 CKB LOC107984648 GPR132 LOC105370687 TDRD9 LOC101929634 C14orf79 BTBD6 LOC105370691 ZBTB42 LINC00638 ZFYVE21 ASPG LOC388022 KIF26A CEP170B AKT1 LOC102723354 MIR203A TRMT61A JAG2 MIR6765 INF2 MIR4710 BAG5 XRCC3 CDCA4 MIR203B ADSSL1 NUDT14 PLD4 APOPT1 TMEM179 LINC00637 PPP1R13B MARK3 LOC105370708 EIF5 PACS2 KLC1 C14orf180 SIVA1 LOC105378183 AHNAK2 BRF1 |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | RD3L XRCC3 C14orf2 LOC107984648 LINC00637 PPP1R13B TDRD9 KLC1 ZFYVE21 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |