Assay Details
Target Gene Details
Entrez Gene ID: | 1755 |
Gene Name: | deleted in malignant brain tumors 1 |
Gene Aliases: |
GP340, SAG, muclin |
Location: |
Chr.10:122560665-122643736 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv7582 | Chr.10:122575775 - 122587364 on Build GRCh38 | Insertion |
![]() |
esv2759786 | Chr.10:122503559 - 122695299 on Build GRCh38 | Gain+Loss |
![]() ![]() |
esv3624775 | Chr.10:122576496 - 122579687 on Build GRCh38 | Loss |
![]() |
esv2760157 | Chr.10:122574497 - 122591497 on Build GRCh38 | Loss |
![]() |
nsv1159772 | Chr.10:122548664 - 122597296 on Build GRCh38 | Duplication |
![]() |
esv3579179 | Chr.10:122575052 - 122597392 on Build GRCh38 | Loss |
![]() |
nsv508611 | Chr.10:122562819 - 122630985 on Build GRCh38 | Deletion |
![]() |
nsv552255 | Chr.10:122522412 - 122617922 on Build GRCh38 | Loss |
![]() |
esv23096 | Chr.10:122546305 - 122617308 on Build GRCh38 | Gain+Loss |
![]() |
esv3577793 | Chr.10:122574679 - 122598197 on Build GRCh38 | Gain |
![]() |
nsv1159773 | Chr.10:122564250 - 122629780 on Build GRCh38 | Duplication |
![]() |
nsv7581 | Chr.10:122569936 - 122639333 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)