Assay Details
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | 4 | 470 | NP_001093588.1 |
NM_001100119.1 | 5 | 564 | NP_001093589.1 | |
NM_005432.3 | 5 | 527 | NP_005423.1 | |
XM_005268046.2 | 4 | 564 | XP_005268103.1 | |
XM_011537138.2 | 5 | 510 | XP_011535440.1 | |
AF035586.1 | 4 | 455 | AAC05368.1 | |
AK022829.1 | 5 | 456 | ||
AK023646.1 | 5 | 500 | ||
AK124498.1 | 3 | 944 | ||
AK126706.1 | 1 | 290 | ||
BC001036.1 | 5 | 499 | AAH01036.1 | |
BC011725.2 | 4 | 430 | AAH11725.1 | |
BT007417.1 | 2 | 147 | AAP36085.1 | |
BX161398.1 | 4 | 439 | CAD61884.1 | |
DB041321.1 | 4 | 487 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | KLC1 LOC107984648 XRCC3 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | KLC1 LOC107984648 XRCC3 ZFYVE21 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | KLC1 LOC107984648 XRCC3 ZFYVE21 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | KLC1 LOC107984648 XRCC3 ZFYVE21 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | CKB APOPT1 LINC00638 C14orf2 INF2 LOC105378183 MIR4710 LOC107984648 LOC102723354 ASPG PLD4 C14orf79 CEP170B PACS2 BAG5 ZBTB42 SIVA1 MIR6765 MIR203A JAG2 GPR132 LINC00637 KLC1 TMEM179 CDCA4 LOC105370687 TDRD9 LOC101929634 EIF5 ZFYVE21 TRMT61A KIF26A MARK3 LOC388022 NUDT14 MIR203B RD3L AKT1 SNORA28 ADSSL1 PPP1R13B LOC105370691 C14orf180 XRCC3 AHNAK2 BTBD6 BRF1 LOC105370708 |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | LINC00637 KLC1 RD3L C14orf2 PPP1R13B TDRD9 LOC107984648 XRCC3 ZFYVE21 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |