Assay Details
Target Gene Details
Entrez Gene ID: | 7306 |
Gene Name: | tyrosinase related protein 1 |
Gene Aliases: |
CAS2, CATB, GP75, OCA3, TRP, TRP1, TYRP, b-PROTEIN |
Location: |
Chr.9:12693385-12710266 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TYRP1 | NM_000550.2 | 3 | 640 | NP_000541.1 |
BC052608.1 | 2 | 559 | AAH52608.1 | |
CR407683.1 | 2 | 462 | CAG28611.1 | |
X51420.1 | 3 | 591 | CAA35785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891612 | Chr.9:12213879 - 13226946 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
dgv12568n54 | Chr.9:12628213 - 12712157 on Build GRCh38 | Loss |
![]() ![]() |
nsv1021844 | Chr.9:12356017 - 13444976 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1016570 | Chr.9:12576095 - 12722388 on Build GRCh38 | Loss |
![]() ![]() |
nsv831513 | Chr.9:12531673 - 12695747 on Build GRCh38 | Gain |
![]() |
esv3619718 | Chr.9:12621841 - 12766938 on Build GRCh38 | Gain |
![]() ![]() |
esv2422212 | Chr.9:12448789 - 13127739 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() |
esv2759671 | Chr.9:12235048 - 12784093 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv613613 | Chr.9:12641987 - 12773264 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)