Assay Details
Target Gene Details
Entrez Gene ID: | 5521 |
Gene Name: | protein phosphatase 2 regulatory subunit Bbeta |
Gene Aliases: |
B55BETA, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55-BETA, PR55BETA, SCA12 |
Location: |
Chr.5:146589504-147081520 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP2R2B | NM_001271899.1 | NP_001258828.1 | ||
NM_001271900.1 | NP_001258829.1 | |||
NM_001271948.1 | NP_001258877.1 | |||
NM_181674.2 | NP_858060.2 | |||
NM_181675.3 | NP_858061.2 | |||
NM_181676.2 | NP_858062.1 | |||
NM_181677.2 | NP_858063.1 | |||
NM_181678.2 | NP_858064.1 | |||
NR_073526.1 | ||||
NR_073527.1 | ||||
AK056192.1 | ||||
AK289717.1 | ||||
AK294659.1 | ||||
AK295347.1 | ||||
AK314810.1 | ||||
BC031790.1 | AAH31790.1 | |||
BX647887.1 | ||||
M64930.1 | AAA36491.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |