Assay Details
Target Gene Details
Entrez Gene ID: | 1201 |
Gene Name: | CLN3, battenin |
Gene Aliases: |
BTS, JNCL |
Location: |
Chr.16:28466653-28492302 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | SGF29 NUPR1 SULT1A1 SULT1A2 IL27 APOBR CLN3 |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | XPO6 NPIP SULT1A1 SULT1A2 CLN3 RABEP2 ATXN2L MIR6862-2 MIR4721 MIR4517 NPIPB6 NFATC2IP EIF3CL NPIPB9 SGF29 MIR6862-1 LOC101928215 CD19 LAT APOBR SBK1 SPNS1 EIF3C NUPR1 SH2B1 IL27 RRN3P2 ATP2A1 NPIPB8 TUFM ATP2A1-AS1 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | NPIP SULT1A1 SULT1A2 CLN3 RABEP2 ATXN2L MIR6862-2 MIR4721 MIR4517 NFATC2IP EIF3CL NPIPB9 SGF29 MIR6862-1 LOC101928215 CD19 LAT APOBR LOC107984832 SPNS1 EIF3C NUPR1 SH2B1 IL27 RRN3P2 ATP2A1 NPIPB8 TUFM ATP2A1-AS1 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | NPIPB6 EIF3CL SGF29 MIR6862-1 NUPR1 IL27 APOBR CLN3 |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | APOBR CLN3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |