Assay Details
Target Gene Details
Entrez Gene ID: | 1890 |
Gene Name: | thymidine phosphorylase |
Gene Aliases: |
ECGF, ECGF1, MEDPS1, MNGIE, MTDPS1, PDECGF, TP, hPD-ECGF |
Location: |
Chr.22:50525752-50530085 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TYMP | NM_001113755.2 | NP_001107227.1 | ||
NM_001113756.2 | NP_001107228.1 | |||
NM_001257988.1 | NP_001244917.1 | |||
NM_001257989.1 | NP_001244918.1 | |||
NM_001953.4 | NP_001944.1 | |||
AK225269.1 | ||||
AK301194.1 | ||||
AK314716.1 | ||||
BC018160.1 | AAH18160.1 | |||
BC052211.2 | AAH52211.1 | |||
KU178037.1 | ||||
KU178038.1 | ||||
KU178039.1 | ||||
M63193.1 | AAA60043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758852 | Chr.22:50491090 - 50578979 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160782 | Chr.22:50463961 - 50571524 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834238 | Chr.22:50402690 - 50556965 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471224 | Chr.22:50435189 - 50580482 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829340 | Chr.22:50505720 - 50603696 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv955201 | Chr.22:50421672 - 50537371 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834239 | Chr.22:50485317 - 50630215 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv589222 | Chr.22:50491152 - 50589773 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)