Assay Details
Target Gene Details
Entrez Gene ID: | 374291 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S7 |
Gene Aliases: |
CI-20, CI-20KD, MY017, PSST |
Location: |
Chr.19:1383571-1395589 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS7 | NM_024407.4 | NP_077718.3 | ||
XM_005259556.4 | XP_005259613.2 | |||
XM_017026768.1 | 4 | 603 | XP_016882257.1 | |
AF060512.1 | AAG43130.1 | |||
AF115969.1 | AAP97240.1 | |||
AK091024.1 | ||||
AK091623.1 | ||||
AK092169.1 | BAC03820.1 | |||
AK127759.1 | 4 | 270 | BAC87117.1 | |
AK128713.1 | BAC87587.1 | |||
AK222738.1 | BAD96458.1 | |||
AK289671.1 | ||||
AK293917.1 | ||||
AK297611.1 | ||||
BC001715.3 | AAH01715.2 | |||
BC005954.1 | AAH05954.1 | |||
BC111517.1 | AAI11518.1 | |||
DC340464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055164 | Chr.19:1246647 - 1389804 on Build GRCh38 | Gain | CIRBP-AS1 MUM1 C19orf24 NDUFS7 CIRBP EFNA2 MIDN |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | ONECUT3 REXO1 CSNK1G2 GAMT PCSK4 BTBD2 C19orf25 PLK5 LOC100288123 UQCR11 SCAMP4 APC2 MIR1909 CSNK1G2-AS1 KLF16 ADAT3 REEP6 MBD3 DAZAP1 ADAMTSL5 ATP8B3 MUM1 RPS15 MEX3D NDUFS7 TCF3 ABHD17A |
nsv833705 | Chr.19:1380125 - 1544934 on Build GRCh38 | Loss | APC2 RPS15 NDUFS7 GAMT REEP6 C19orf25 PLK5 DAZAP1 ADAMTSL5 PCSK4 |
nsv482207 | Chr.19:1383884 - 1395589 on Build GRCh38 | Gain | NDUFS7 |
esv3643389 | Chr.19:1264454 - 1397379 on Build GRCh38 | Gain | CIRBP-AS1 MUM1 C19orf24 NDUFS7 GAMT CIRBP EFNA2 |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | WDR18 GAMT CBARP PCSK4 LOC102725180 CIRBP-AS1 POLR2E ATP5D ARHGAP45 GRIN3B C19orf25 PLK5 CIRBP STK11 APC2 RNU6-2 CNN2 SBNO2 REEP6 TMEM259 DAZAP1 ADAMTSL5 GPX4 ARID3A MUM1 C19orf24 RPS15 NDUFS7 LOC105372235 EFNA2 MIDN ABCA7 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | ONECUT3 REXO1 CSNK1G2 GAMT PCSK4 BTBD2 C19orf25 PLK5 LOC100288123 UQCR11 SCAMP4 APC2 MIR1909 CSNK1G2-AS1 MKNK2 KLF16 ADAT3 REEP6 MBD3 DAZAP1 ADAMTSL5 ATP8B3 MUM1 RPS15 MEX3D NDUFS7 TCF3 ABHD17A |
esv3643396 | Chr.19:1356856 - 1396468 on Build GRCh38 | Gain | MUM1 NDUFS7 |
nsv578278 | Chr.19:1371355 - 1448181 on Build GRCh38 | Loss | APC2 MUM1 RPS15 NDUFS7 GAMT DAZAP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |