Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC1 | NM_001130107.1 | NP_001123579.1 | ||
NM_182923.3 | NP_891553.2 | |||
AB451299.1 | ||||
AB451431.1 | ||||
AY180165.3 | AAO62550.1 | |||
AY180168.1 | AAO62553.1 | |||
BM988840.1 | ||||
L04733.1 | AAA16576.1 |
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | ||||
BC001036.1 | AAH01036.1 | |||
BC002949.1 | AAH02949.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | LOC107984648 XRCC3 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | LOC107984648 ZFYVE21 XRCC3 KLC1 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | LOC107984648 ZFYVE21 XRCC3 KLC1 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | LOC107984648 ZFYVE21 XRCC3 KLC1 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | CDCA4 LINC00637 C14orf180 INF2 MIR4710 APOPT1 RD3L ZFYVE21 XRCC3 SNORA28 LOC102723354 NUDT14 BTBD6 LOC101929634 LINC00638 TDRD9 AHNAK2 C14orf2 BRF1 EIF5 LOC107984648 TMEM179 AKT1 ASPG PPP1R13B MIR203B KIF26A SIVA1 LOC105370687 BAG5 C14orf79 LOC105378183 MIR203A ADSSL1 CEP170B GPR132 JAG2 TRMT61A ZBTB42 PACS2 LOC105370708 LOC105370691 LOC388022 MIR6765 PLD4 KLC1 MARK3 CKB |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | LINC00637 PPP1R13B TDRD9 C14orf2 LOC107984648 RD3L ZFYVE21 XRCC3 KLC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |