Assay Details
Target Gene Details
Entrez Gene ID: | 1201 |
Gene Name: | CLN3, battenin |
Gene Aliases: |
BTS, JNCL |
Location: |
Chr.16:28466653-28492302 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | SULT1A1 CLN3 NUPR1 SULT1A2 APOBR IL27 SGF29 |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | NUPR1 RABEP2 EIF3C MIR6862-2 MIR4517 NPIPB6 NFATC2IP LAT EIF3CL SULT1A2 XPO6 MIR6862-1 MIR4721 ATXN2L ATP2A1 CLN3 ATP2A1-AS1 IL27 NPIPB8 SGF29 LOC101928215 SULT1A1 NPIP RRN3P2 CD19 TUFM APOBR SBK1 SH2B1 SPNS1 NPIPB9 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | NUPR1 RABEP2 EIF3C MIR6862-2 MIR4517 NFATC2IP LAT EIF3CL SULT1A2 MIR6862-1 MIR4721 ATXN2L ATP2A1 CLN3 ATP2A1-AS1 IL27 NPIPB8 SGF29 LOC101928215 SULT1A1 NPIP RRN3P2 CD19 LOC107984832 TUFM APOBR SH2B1 SPNS1 NPIPB9 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | CLN3 NUPR1 EIF3CL MIR6862-1 APOBR NPIPB6 IL27 SGF29 |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | CLN3 APOBR |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |