Assay Details
Target Gene Details
Entrez Gene ID: | 1649 |
Gene Name: | DNA damage inducible transcript 3 |
Gene Aliases: |
CEBPZ, CHOP, CHOP-10, CHOP10, GADD153 |
Location: |
Chr.12:57516588-57520517 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDIT3 | NM_001195053.1 | NP_001181982.1 | ||
NM_001195054.1 | NP_001181983.1 | |||
NM_001195055.1 | NP_001181984.1 | |||
NM_001195056.1 | NP_001181985.1 | |||
NM_001195057.1 | NP_001181986.1 | |||
NM_004083.5 | NP_004074.2 | |||
AA054097.1 | ||||
AA476561.1 | ||||
AK316581.1 | ||||
AV729744.1 | ||||
AY461580.1 | ||||
BC003637.2 | AAH03637.1 | |||
BM464290.1 | ||||
BU664077.1 | ||||
CN482428.1 | ||||
S40706.1 | AAB22646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain | MIR6758 MIR616 DDIT3 DTX3 ARHGAP9 PIP4K2C INHBE KIF5A DCTN2 GLI1 MBD6 INHBC MARS |
nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss | MIR6758 MIR616 DDIT3 DTX3 ARHGAP9 LOC101927583 PIP4K2C KIF5A DCTN2 GLI1 ARHGEF25 MBD6 MARS |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss | MIR6758 INHBE MIR616 DDIT3 GLI1 MBD6 ARHGAP9 INHBC MARS |
nsv559036 | Chr.12:57517429 - 57522259 on Build GRCh38 | Loss | MIR616 DDIT3 MBD6 |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | MIR616 ARHGAP9 LOC101927583 NABP2 SNORD59A CNPY2 SLC39A5 RDH16 OS9 GLS2 NEMP1 MIP ZBTB39 TIMELESS TAC3 BAZ2A SPRYD4 PAN2 COQ10A IL23A KIF5A CS GPR182 PTGES3 NACA STAT6 LRP1-AS MIR6758 B4GALNT1 DDIT3 STAC3 MYO1A LRP1 HSD17B6 SNORD59B PIP4K2C SLC26A10 INHBE NAB2 DCTN2 SNORA105C MBD6 SHMT2 NDUFA4L2 HBCBP ANKRD52 RBMS2 MIR1228 DTX3 RNF41 PRIM1 ATP5B R3HDM2 SDR9C7 NXPH4 GLI1 ARHGEF25 INHBC STAT2 MARS APOF |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |