Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC1 | NM_001130107.1 | NP_001123579.1 | ||
NM_182923.3 | NP_891553.2 | |||
AB451299.1 | ||||
AB451431.1 | ||||
AY180165.3 | AAO62550.1 | |||
AY180168.1 | AAO62553.1 | |||
BM988840.1 | ||||
L04733.1 | AAA16576.1 |
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | ||||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | LOC107984648 XRCC3 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | LOC107984648 XRCC3 KLC1 ZFYVE21 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | LOC107984648 XRCC3 KLC1 ZFYVE21 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | LOC107984648 XRCC3 KLC1 ZFYVE21 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | JAG2 LOC107984648 GPR132 BAG5 ZFYVE21 BTBD6 KIF26A LOC105370687 CDCA4 CKB ZBTB42 C14orf180 PPP1R13B INF2 LOC102723354 ASPG SIVA1 LOC105370691 PACS2 ADSSL1 LINC00638 PLD4 C14orf79 MIR203B APOPT1 TMEM179 MIR4710 C14orf2 BRF1 NUDT14 AHNAK2 MIR6765 LOC101929634 MIR203A LINC00637 LOC388022 AKT1 MARK3 XRCC3 KLC1 TDRD9 SNORA28 TRMT61A EIF5 LOC105378183 LOC105370708 CEP170B RD3L |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | PPP1R13B LOC107984648 C14orf2 XRCC3 KLC1 TDRD9 LINC00637 ZFYVE21 RD3L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |