Assay Details
Target Gene Details
Entrez Gene ID: | 1201 |
Gene Name: | CLN3, battenin |
Gene Aliases: |
BTS, JNCL |
Location: |
Chr.16:28466653-28492302 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | SULT1A1 NUPR1 CLN3 IL27 SGF29 SULT1A2 APOBR |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | SULT1A1 ATP2A1-AS1 IL27 SH2B1 SPNS1 RABEP2 EIF3CL NPIPB9 XPO6 NUPR1 TUFM MIR4721 MIR6862-2 ATXN2L LAT NPIPB8 RRN3P2 MIR6862-1 LOC101928215 SGF29 SULT1A2 NPIP SBK1 APOBR MIR4517 CLN3 NPIPB6 NFATC2IP EIF3C ATP2A1 CD19 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | SULT1A1 ATP2A1-AS1 IL27 SH2B1 SPNS1 RABEP2 EIF3CL NPIPB9 NUPR1 TUFM MIR4721 MIR6862-2 ATXN2L LOC107984832 LAT NPIPB8 RRN3P2 MIR6862-1 LOC101928215 SGF29 SULT1A2 NPIP APOBR MIR4517 CLN3 NFATC2IP EIF3C ATP2A1 CD19 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | EIF3CL NUPR1 CLN3 NPIPB6 IL27 MIR6862-1 SGF29 APOBR |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | CLN3 APOBR |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |