Assay Details
Target Gene Details
Entrez Gene ID: | 374291 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S7 |
Gene Aliases: |
CI-20, CI-20KD, MY017, PSST |
Location: |
Chr.19:1383571-1395589 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS7 | NM_024407.4 | NP_077718.3 | ||
XM_005259556.4 | XP_005259613.2 | |||
XM_017026768.1 | XP_016882257.1 | |||
AF060512.1 | AAG43130.1 | |||
AF115969.1 | AAP97240.1 | |||
AK091024.1 | ||||
AK091623.1 | ||||
AK092169.1 | 3 | 168 | BAC03820.1 | |
AK127759.1 | BAC87117.1 | |||
AK128713.1 | 3 | 367 | BAC87587.1 | |
AK222738.1 | BAD96458.1 | |||
AK289671.1 | ||||
AK293917.1 | ||||
AK297611.1 | ||||
BC001715.3 | AAH01715.2 | |||
BC005954.1 | AAH05954.1 | |||
BC111517.1 | AAI11518.1 | |||
DC340464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055164 | Chr.19:1246647 - 1389804 on Build GRCh38 | Gain | CIRBP-AS1 EFNA2 MIDN C19orf24 NDUFS7 MUM1 CIRBP |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | LOC100288123 NDUFS7 MBD3 MEX3D ATP8B3 KLF16 MUM1 UQCR11 APC2 ABHD17A RPS15 PCSK4 ONECUT3 GAMT MIR1909 CSNK1G2 PLK5 SCAMP4 REXO1 ADAT3 REEP6 TCF3 DAZAP1 ADAMTSL5 CSNK1G2-AS1 BTBD2 C19orf25 |
nsv833705 | Chr.19:1380125 - 1544934 on Build GRCh38 | Loss | RPS15 PCSK4 PLK5 NDUFS7 REEP6 DAZAP1 ADAMTSL5 GAMT C19orf25 APC2 |
nsv482207 | Chr.19:1383884 - 1395589 on Build GRCh38 | Gain | NDUFS7 |
esv3643389 | Chr.19:1264454 - 1397379 on Build GRCh38 | Gain | CIRBP-AS1 EFNA2 C19orf24 NDUFS7 MUM1 GAMT CIRBP |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | STK11 NDUFS7 GPX4 MUM1 CBARP APC2 LOC102725180 POLR2E RPS15 PCSK4 GRIN3B CNN2 GAMT SBNO2 ABCA7 CIRBP-AS1 TMEM259 WDR18 PLK5 MIDN C19orf24 REEP6 RNU6-2 DAZAP1 ADAMTSL5 CIRBP ARID3A EFNA2 LOC105372235 ATP5D C19orf25 ARHGAP45 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | LOC100288123 MKNK2 NDUFS7 MBD3 MEX3D ATP8B3 KLF16 MUM1 UQCR11 APC2 ABHD17A RPS15 PCSK4 ONECUT3 GAMT MIR1909 CSNK1G2 PLK5 SCAMP4 REXO1 ADAT3 REEP6 TCF3 DAZAP1 ADAMTSL5 CSNK1G2-AS1 BTBD2 C19orf25 |
esv3643396 | Chr.19:1356856 - 1396468 on Build GRCh38 | Gain | NDUFS7 MUM1 |
nsv578278 | Chr.19:1371355 - 1448181 on Build GRCh38 | Loss | RPS15 NDUFS7 DAZAP1 MUM1 GAMT APC2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |