Assay Details
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | ||||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | XRCC3 KLC1 LOC107984648 |
esv2749175 | Chr.14:103700180 - 103705264 on Build GRCh38 | Deletion | XRCC3 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | ZFYVE21 XRCC3 KLC1 LOC107984648 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | ZFYVE21 XRCC3 KLC1 LOC107984648 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | ZFYVE21 XRCC3 KLC1 LOC107984648 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | MIR6765 MIR4710 NUDT14 MIR203A XRCC3 INF2 CEP170B RD3L PLD4 TDRD9 LOC105378183 LOC101929634 LOC105370708 PACS2 ZFYVE21 EIF5 LINC00637 GPR132 SNORA28 C14orf180 JAG2 ADSSL1 C14orf79 KIF26A AKT1 LOC105370691 PPP1R13B C14orf2 AHNAK2 BTBD6 BAG5 MARK3 LOC102723354 LOC388022 TMEM179 LOC107984648 ASPG ZBTB42 CDCA4 SIVA1 LINC00638 BRF1 LOC105370687 CKB KLC1 MIR203B APOPT1 TRMT61A |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | ZFYVE21 PPP1R13B C14orf2 LINC00637 XRCC3 KLC1 LOC107984648 RD3L TDRD9 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |