Assay Details
Target Gene Details
Entrez Gene ID: | 374291 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S7 |
Gene Aliases: |
CI-20, CI-20KD, MY017, PSST |
Location: |
Chr.19:1383571-1395589 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS7 | NM_024407.4 | NP_077718.3 | ||
XM_005259556.4 | XP_005259613.2 | |||
AF060512.1 | AAG43130.1 | |||
AF115969.1 | AAP97240.1 | |||
AK091024.1 | ||||
AK091046.1 | 1 | 1418 | ||
AK091623.1 | ||||
AK092169.1 | BAC03820.1 | |||
AK127759.1 | BAC87117.1 | |||
AK128713.1 | BAC87587.1 | |||
AK222738.1 | BAD96458.1 | |||
AK289671.1 | ||||
AK293917.1 | ||||
AK297611.1 | ||||
BC001715.3 | AAH01715.2 | |||
BC005954.1 | AAH05954.1 | |||
BC111517.1 | AAI11518.1 | |||
DC340464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | ONECUT3 GAMT ADAMTSL5 ABHD17A UQCR11 MUM1 ATP8B3 CSNK1G2-AS1 MIR1909 APC2 TCF3 MBD3 MEX3D NDUFS7 SCAMP4 C19orf25 RPS15 REEP6 REXO1 ADAT3 BTBD2 DAZAP1 PCSK4 PLK5 KLF16 LOC100288123 CSNK1G2 |
nsv833705 | Chr.19:1380125 - 1544934 on Build GRCh38 | Loss | GAMT ADAMTSL5 DAZAP1 NDUFS7 PCSK4 PLK5 C19orf25 RPS15 REEP6 APC2 |
nsv482207 | Chr.19:1383884 - 1395589 on Build GRCh38 | Gain | NDUFS7 |
esv3643389 | Chr.19:1264454 - 1397379 on Build GRCh38 | Gain | GAMT EFNA2 C19orf24 NDUFS7 MUM1 CIRBP-AS1 CIRBP |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | GRIN3B TMEM259 GAMT ADAMTSL5 CBARP MUM1 MIDN CIRBP APC2 POLR2E CNN2 C19orf24 ARID3A GPX4 SBNO2 ATP5D NDUFS7 LOC102725180 C19orf25 ABCA7 ARHGAP45 RPS15 CIRBP-AS1 REEP6 EFNA2 RNU6-2 DAZAP1 LOC105372235 STK11 PCSK4 PLK5 WDR18 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | ONECUT3 GAMT ADAMTSL5 ABHD17A UQCR11 MUM1 ATP8B3 CSNK1G2-AS1 MIR1909 APC2 TCF3 MBD3 MEX3D NDUFS7 SCAMP4 C19orf25 RPS15 REEP6 REXO1 ADAT3 MKNK2 BTBD2 DAZAP1 PCSK4 PLK5 KLF16 LOC100288123 CSNK1G2 |
esv3643396 | Chr.19:1356856 - 1396468 on Build GRCh38 | Gain | NDUFS7 MUM1 |
nsv578278 | Chr.19:1371355 - 1448181 on Build GRCh38 | Loss | GAMT DAZAP1 NDUFS7 MUM1 RPS15 APC2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |